COM448635527 Combi-Blocks, Inc.

5-Fluoro-2-morpholinopyridine

  • MDL number MFCD18089238
  • Cas Number : 1287217-51-9
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1877
Availability details for
Quantity
  • Availability
  • Add to Cart