COM448635529 Combi-Blocks, Inc.

5-Fluoro-2-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD18089240
  • Cas Number : 1287217-79-1
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1879
Availability details for
Quantity
  • Availability
  • Add to Cart