COM448635532 Combi-Blocks, Inc.

2-Acetamido-6-trifluoromethylpyridine

  • MDL number MFCD18837632
  • Cas Number : 1291487-27-8
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1894
Availability details for
Quantity
  • Availability
  • Add to Cart