COM448635534 Combi-Blocks, Inc.

2-Amino-3,5-dibromo-6-trifluoropyridine

  • MDL number MFCD18837633
  • Cas Number : 1291487-16-5
  • Molecular Weight : 319.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1898
Availability details for
Quantity
  • Availability
  • Add to Cart