COM448635537 Combi-Blocks, Inc.

2-(N,N-DiBOC-Amino)-5-bromopyridine

  • MDL number MFCD18249958
  • Cas Number : 209959-28-4
  • Molecular Weight : 373.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1902
Availability details for
Quantity
  • Availability
  • Add to Cart