COM448635546 Combi-Blocks, Inc.

2-tert-Butoxy-p-picoline

  • MDL number MFCD18914537
  • Cas Number : 57883-15-5
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1911
Availability details for
Quantity
  • Availability
  • Add to Cart