COM448635547 Combi-Blocks, Inc.

2-Methoxy-6-p-tolylpyridine

  • MDL number MFCD18914538
  • Cas Number : 1039775-38-6
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1912
Availability details for
Quantity
  • Availability
  • Add to Cart