COM448635548 Combi-Blocks, Inc.

2-Chloro-5-(trifluoromethyl)pyridin-3-ol

  • MDL number MFCD13189916
  • Cas Number : 1196153-98-6
  • Molecular Weight : 197.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1913

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1913
Availability details for
Quantity
  • Availability
  • Add to Cart