COM448635551 Combi-Blocks, Inc.

2-Methoxy-6-(4-(trifluoromethyl)phenyl)pyridine

  • MDL number MFCD18421599
  • Cas Number : 1202026-99-0
  • Molecular Weight : 253.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1918
Availability details for
Quantity
  • Availability
  • Add to Cart