COM448635552 Combi-Blocks, Inc.

2,3-Dichloro-5-methoxypyridine

  • MDL number MFCD08277295
  • Cas Number : 885168-12-7
  • Molecular Weight : 178.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1919
Availability details for
Quantity
  • Availability
  • Add to Cart