COM448635553 Combi-Blocks, Inc.

1-(5-Bromopyridin-2-yl)piperidine-3-carboxylic acid

  • MDL number MFCD09739754
  • Cas Number : 954267-45-9
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1920
Availability details for
Quantity
  • Availability
  • Add to Cart