COM448635559 Combi-Blocks, Inc.

Ethyl 2-thiouracil-5-carboxylate

  • MDL number MFCD01080781
  • Cas Number : 38026-46-9
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1927
Availability details for
Quantity
  • Availability
  • Add to Cart