COM448635560 Combi-Blocks, Inc.

2-(2-Bromophenoxymethyl)pyridine

  • MDL number MFCD09927519
  • Cas Number : 1016531-68-2
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1929
Availability details for
Quantity
  • Availability
  • Add to Cart