COM448635561 Combi-Blocks, Inc.

2-(3-Bromophenoxymethyl)pyridine

  • MDL number MFCD09928322
  • Cas Number : 488799-65-1
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1933
Availability details for
Quantity
  • Availability
  • Add to Cart