COM448635563 Combi-Blocks, Inc.

4-fluoropyridine-2-carboxylic acid

  • MDL number MFCD07374895
  • Cas Number : 886371-78-4
  • Molecular Weight : 141.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1937
Availability details for
Quantity
  • Availability
  • Add to Cart