COM448635577 Combi-Blocks, Inc.

7-Bromo-[1,2,3,4]tetrazolo[1,5-a]pyridine

  • MDL number MFCD19684133
  • Cas Number : 1130816-25-9
  • Molecular Weight : 199.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1957
Availability details for
Quantity
  • Availability
  • Add to Cart