COM448635582 Combi-Blocks, Inc.

5-(Benzyloxy)-2,3-dichloropyridine

  • MDL number MFCD19684134
  • Cas Number : 1314987-39-7
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1963

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1963
Availability details for
Quantity
  • Availability
  • Add to Cart