COM448635587 Combi-Blocks, Inc.

2-Methoxy-6-methyl-3-nitropyridine

  • MDL number MFCD03095074
  • Cas Number : 112163-03-8
  • Molecular Weight : 168.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1970
Availability details for
Quantity
  • Availability
  • Add to Cart