COM448635599 Combi-Blocks, Inc.

1-(2-methylpyrimidin-5-yl)ethanone

  • MDL number MFCD18073349
  • Cas Number : 1225223-42-6
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-1984
Availability details for
Quantity
  • Availability
  • Add to Cart