COM448635605 Combi-Blocks, Inc.

3-Bromo-2,5-dimethoxypyridine

  • MDL number MFCD26398166
  • Cas Number : 1093951-75-7
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2000
Availability details for
Quantity
  • Availability
  • Add to Cart