COM448635606 Combi-Blocks, Inc.

2-Hydroxy-5-methoxypyridine

  • MDL number MFCD11044353
  • Cas Number : 61941-79-5
  • Molecular Weight : 125.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2002

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2002
Availability details for
Quantity
  • Availability
  • Add to Cart