COM448635607 Combi-Blocks, Inc.

2-Bromo-3,6-dimethoxypyridine

  • MDL number MFCD18255848
  • Cas Number : 1211525-11-9
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-2003
Availability details for
Quantity
  • Availability
  • Add to Cart