COM448635620 Combi-Blocks, Inc.

6-Iodonicotinonitrile

  • MDL number MFCD08689512
  • Cas Number : 289470-22-0
  • Molecular Weight : 230.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7009
Availability details for
Quantity
  • Availability
  • Add to Cart