COM448635628 Combi-Blocks, Inc.

N-Benzyl-4-(benzylamino)picolinamide

  • MDL number MFCD09027296
  • Cas Number : 913836-29-0
  • Molecular Weight : 317.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7020
Availability details for
Quantity
  • Availability
  • Add to Cart