COM448635631 Combi-Blocks, Inc.

2-Methoxyisonicotinamide

  • MDL number MFCD00174337
  • Cas Number : 105612-50-8
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7030
Availability details for
Quantity
  • Availability
  • Add to Cart