COM448635641 Combi-Blocks, Inc.

N,N-Dimethylpyridine-3-sulfonamide

  • MDL number MFCD07368248
  • Cas Number : 4810-41-7
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7045
Availability details for
Quantity
  • Availability
  • Add to Cart