COM448635642 Combi-Blocks, Inc.

N-Ethylpyridine-3-sulfonamide

  • MDL number MFCD09027298
  • Cas Number : 4810-40-6
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7046
Availability details for
Quantity
  • Availability
  • Add to Cart