COM448635654 Combi-Blocks, Inc.

4-(Pyridin-3-ylsulfonyl)morpholine

  • MDL number MFCD00090297
  • Cas Number : 26103-48-0
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7074
Availability details for
Quantity
  • Availability
  • Add to Cart