COM448635666 Combi-Blocks, Inc.

1-(4-Bromophenyl)-3-cyano-2(1H)-pyridinone

  • MDL number MFCD09258784
  • Cas Number : 929000-84-0
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7087
Availability details for
Quantity
  • Availability
  • Add to Cart