COM448635683 Combi-Blocks, Inc.

Ethyl 6-bromo-7-methylimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD09475851
  • Cas Number : 907945-87-3
  • Molecular Weight : 283.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7112
Availability details for
Quantity
  • Availability
  • Add to Cart