COM448635690 Combi-Blocks, Inc.

4-(2-Chloropyridin-3-ylsulfonyl)morpholine

  • MDL number MFCD09475837
  • Cas Number : 60597-72-0
  • Molecular Weight : 262.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7124
Availability details for
Quantity
  • Availability
  • Add to Cart