COM448635723 Combi-Blocks, Inc.

2,6,7-Trichloro-3-trifluoromethylquinoxaline

  • MDL number MFCD09801052
  • Cas Number : 281209-13-0
  • Molecular Weight : 301.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7168
Availability details for
Quantity
  • Availability
  • Add to Cart