COM448635732 Combi-Blocks, Inc.

6-Bromo-8-nitro-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD09801055
  • Cas Number : 951884-20-1
  • Molecular Weight : 243.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7181

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7181
Availability details for
Quantity
  • Availability
  • Add to Cart