COM448635753 Combi-Blocks, Inc.

N-Cyclopentyl 5-bromopicolinamide

  • MDL number MFCD09801064
  • Cas Number : 845305-91-1
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7212

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7212
Availability details for
Quantity
  • Availability
  • Add to Cart