COM448635755 Combi-Blocks, Inc.

5-Bromo-N-butylpicolinamide

  • MDL number MFCD09801065
  • Cas Number : 951885-14-6
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7214
Availability details for
Quantity
  • Availability
  • Add to Cart