COM448635760 Combi-Blocks, Inc.

2-Bromo-N-methylisonicotinamide

  • MDL number MFCD03411643
  • Cas Number : 337536-01-3
  • Molecular Weight : 215.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7227
Availability details for
Quantity
  • Availability
  • Add to Cart