COM448635762 Combi-Blocks, Inc.

2-Bromo-N-cyclopropylisonicotinamide

  • MDL number MFCD09878430
  • Cas Number : 951885-74-8
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7229
Availability details for
Quantity
  • Availability
  • Add to Cart