COM448635767 Combi-Blocks, Inc.

6,7-Dichloroquinoline

  • MDL number MFCD09878431
  • Cas Number : 40635-11-8
  • Molecular Weight : 198.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7236
Availability details for
Quantity
  • Availability
  • Add to Cart