COM448635782 Combi-Blocks, Inc.

3-Bromo-5-(4-methoxybenzyloxy)pyridine

  • MDL number MFCD09878436
  • Cas Number : 552331-73-4
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7254
Availability details for
Quantity
  • Availability
  • Add to Cart