COM448635794 Combi-Blocks, Inc.

Ethyl 4-ethoxynicotinate

  • MDL number MFCD09972185
  • Cas Number : 33279-63-9
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7269
Availability details for
Quantity
  • Availability
  • Add to Cart