COM448635812 Combi-Blocks, Inc.

5-Aminoisoquinoline, HCl

  • MDL number MFCD00035269
  • Cas Number : 152814-23-8
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7289

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7289
Availability details for
Quantity
  • Availability
  • Add to Cart