COM448635814 Combi-Blocks, Inc.

4-(5-Chloro-3-fluoro-2-pyridinyl)morpholine

  • MDL number MFCD09972191
  • Cas Number : 1020253-17-1
  • Molecular Weight : 216.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7297
Availability details for
Quantity
  • Availability
  • Add to Cart