COM448635827 Combi-Blocks, Inc.

5-Chloro-3-fluoro-2-(N-propylamino)pyridine, HCl

  • MDL number MFCD09972199
  • Cas Number : 1073372-11-8
  • Molecular Weight : 225.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7310
Availability details for
Quantity
  • Availability
  • Add to Cart