COM448635833 Combi-Blocks, Inc.

2-Methylamino-5-methyl-3-nitropyridine

  • MDL number MFCD09862683
  • Cas Number : 106690-38-4
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7318
Availability details for
Quantity
  • Availability
  • Add to Cart