COM448635834 Combi-Blocks, Inc.

2-Cyclopentylamino-5-methyl-3-nitropyridine

  • MDL number MFCD10699704
  • Cas Number : 1033202-29-7
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7319
Availability details for
Quantity
  • Availability
  • Add to Cart