COM448635835 Combi-Blocks, Inc.

2-Hydroxy-4-methyl-6-(trifluoromethyl)nicotinonitrile

  • MDL number MFCD04967406
  • Cas Number : 116548-08-4
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7320
Availability details for
Quantity
  • Availability
  • Add to Cart