COM448635845 Combi-Blocks, Inc.

5-Bromo-N-(4-methoxybenzyl)-3-nitropyridin-2-amine

  • MDL number MFCD10699706
  • Cas Number : 1033202-35-5
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7333
Availability details for
Quantity
  • Availability
  • Add to Cart