COM448635849 Combi-Blocks, Inc.

2-Chloro-6-(4-methoxybenzyloxy)pyridine

  • MDL number MFCD09972200
  • Cas Number : 1020253-23-9
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7338

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7338
Availability details for
Quantity
  • Availability
  • Add to Cart