COM448635852 Combi-Blocks, Inc.

3-Bromo-2-methoxy-5-trifluoromethylpyridine

  • MDL number MFCD09972202
  • Cas Number : 124432-63-9
  • Molecular Weight : 256.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7341
Availability details for
Quantity
  • Availability
  • Add to Cart