COM448635856 Combi-Blocks, Inc.

2,2'-Bipyridine-3,3'-dicarbonitrile

  • MDL number MFCD09972203
  • Cas Number : 136869-49-3
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7345
Availability details for
Quantity
  • Availability
  • Add to Cart